Assay Details
| Gene Symbol: ivd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011380.1 | NP_001011380.1 | 4 - 5 | 465 | 67 | |
| GenBank mRNA | BC088561.1 | - | 4 - 5 | 465 | 67 | |
| CT030360.1 | - | 4 - 5 | 453 | 67 | ||
More Information
Set Membership: |
|

ivd
Genomic Map