Assay Details
| Gene Symbol: thul16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011392.1 | NP_001011392.1 | 11 - 12 | 1552 | 68 | |
| XM_018096243.1 | XP_017951732.1 | 11 - 12 | 1077 | 68 | ||
| XM_018096244.1 | XP_017951733.1 | 11 - 12 | 1319 | 68 | ||
| GenBank mRNA | BC088584.1 | - | 11 - 12 | 1552 | 68 | |
More Information
Set Membership: |
|

thul16
Genomic Map