Assay Details
| Gene Symbol: thul16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011392.1 | NP_001011392.1 | 1 - 2 | 499 | 74 | |
| GenBank mRNA | BC088584.1 | - | 1 - 2 | 499 | 74 | |
More Information
Set Membership: |
|

thul16
Genomic Map