Assay Details
| Gene Symbol: thul16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011392.1 | NP_001011392.1 | 4 - 5 | 743 | 90 | |
| XM_018096243.1 | XP_017951732.1 | 4 - 5 | 268 | 90 | ||
| XM_018096244.1 | XP_017951733.1 | 4 - 5 | 510 | 90 | ||
| GenBank mRNA | BC088584.1 | - | 4 - 5 | 743 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018096243.1, XM_018096244.1 transcripts.
Set Membership: |
|

thul16
Genomic Map