Assay Details
| Gene Symbol: mfsd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011396.1 | NP_001011396.1 | 8 - 9 | 1538 | 57 | |
| XM_018092856.1 | XP_017948345.1 | 8 - 9 | 1602 | 57 | ||
| GenBank mRNA | BC088599.1 | - | 8 - 9 | 1538 | 57 | |
| CR760546.2 | - | 8 - 9 | 1464 | 57 | ||
More Information
Set Membership: |
|

mfsd12
Genomic Map