Assay Details
| Gene Symbol: src | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011413.1 | NP_001011413.1 | 5 - 6 | 708 | 65 | |
| XM_018097722.1 | XP_017953211.1 | 6 - 7 | 582 | 65 | ||
| XM_018097723.1 | XP_017953212.1 | 6 - 7 | 582 | 65 | ||
| XM_018097724.1 | XP_017953213.1 | 5 - 6 | 567 | 65 | ||
| XM_018097725.1 | XP_017953214.1 | 5 - 6 | 566 | 65 | ||
| XM_018097726.1 | XP_017953215.1 | 6 - 7 | 667 | 65 | ||
| XM_018097727.1 | XP_017953216.1 | 6 - 7 | 923 | 65 | ||
| XM_018097728.1 | XP_017953217.1 | 5 - 6 | 907 | 65 | ||
| GenBank mRNA | AY842261.1 | - | 5 - 6 | 708 | 65 | |
More Information
Set Membership: |
|

src
Genomic Map