Assay Details
| Gene Symbol: myoz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011415.1 | NP_001011415.1 | 5 - 6 | 628 | 79 | |
| XM_012961302.2 | XP_012816756.1 | 6 - 7 | 1122 | 79 | ||
| XM_012961309.2 | XP_012816763.1 | 6 - 7 | 1120 | 79 | ||
| XM_012961316.2 | XP_012816770.1 | 5 - 6 | 1026 | 79 | ||
| GenBank mRNA | BC087984.1 | - | 5 - 6 | 628 | 79 | |
More Information
Set Membership: |
|

myoz2
Genomic Map