Assay Details
| Gene Symbol: rpl22l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011427.1 | NP_001011427.1 | 2 - 3 | 107 | 104 | |
| GenBank mRNA | BC088059.1 | - | 2 - 3 | 107 | 104 | |
| CT025232.2 | - | 1 - 2 | 103 | 104 | ||
More Information
Set Membership: |
|

rpl22l1
Genomic Map