Assay Details
| Gene Symbol: adh1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011431.1 | NP_001011431.1 | 4 - 5 | 385 | 94 | |
| GenBank mRNA | BC088070.1 | - | 4 - 5 | 385 | 94 | |
More Information
Set Membership: |
|

adh1b
Genomic Map