Assay Details
| Gene Symbol: metap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011454.1 | NP_001011454.1 | 4 - 5 | 357 | 69 | |
| XM_018092861.1 | XP_017948350.1 | 4 - 5 | 518 | 69 | ||
| GenBank mRNA | BC088554.1 | - | 4 - 5 | 357 | 69 | |
More Information
Set Membership: |
|

metap1
Genomic Map