Assay Details
| Gene Symbol: alg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011458.1 | NP_001011458.1 | 8 - 9 | 841 | 68 | |
| GenBank mRNA | BC088570.1 | - | 8 - 9 | 841 | 68 | |
| CR761124.2 | - | 8 - 9 | 852 | 68 | ||
More Information
Set Membership: |
|

alg6
Genomic Map