Assay Details
| Gene Symbol: twf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011469.1 | NP_001011469.1 | 2 - 3 | 145 | 77 | |
| GenBank mRNA | BC088597.1 | - | 2 - 3 | 145 | 77 | |
More Information
Set Membership: |
|

twf1
Genomic Map