Assay Details
| Gene Symbol: gnai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC088604.1 | - | 8 - 9 | 1098 | 81 | |
| CR942680.1 | - | 8 - 9 | 1095 | 81 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gnai3| Gene Symbol: gnai3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC088604.1 | - | 8 - 9 | 1098 | 81 | |
| CR942680.1 | - | 8 - 9 | 1095 | 81 | ||
Set Membership: |
|