Assay Details
| Gene Symbol: bud23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011475.1 | NP_001011475.1 | 8 - 9 | 598 | 71 | |
| XM_012956880.1 | XP_012812334.1 | 8 - 9 | 585 | 71 | ||
| XM_018091217.1 | XP_017946706.1 | 6 - 7 | 448 | 71 | ||
| GenBank mRNA | BC088766.1 | - | 8 - 9 | 598 | 71 | |
| BC159372.1 | - | 7 - 8 | 431 | 71 | ||
More Information
Set Membership: |
|

bud23
Genomic Map