Assay Details
| Gene Symbol: ndufv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011491.1 | NP_001011491.1 | 9 - 10 | 1258 | 71 | |
| XM_012961018.2 | XP_012816472.1 | 10 - 11 | 1448 | 71 | ||
| XM_012961019.2 | XP_012816473.1 | 9 - 10 | 1307 | 71 | ||
| XM_012961020.2 | XP_012816474.1 | 8 - 9 | 1194 | 71 | ||
| XM_018095376.1 | XP_017950865.1 | 10 - 11 | 1469 | 71 | ||
| XM_018095377.1 | XP_017950866.1 | 9 - 10 | 1276 | 71 | ||
| GenBank mRNA | BC088876.1 | - | 9 - 10 | 1258 | 71 | |
| CR762342.2 | - | 9 - 10 | 1280 | 71 | ||
More Information
Set Membership: |
|

ndufv1
Genomic Map