Assay Details
| Gene Symbol: crb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012688.1 | NP_001012706.1 | 3 - 4 | 191 | 68 | |
| GenBank mRNA | AY884237.1 | - | 3 - 4 | 191 | 68 | |
More Information
Set Membership: |
|

crb3
Genomic Map