Assay Details
| Gene Symbol: ugdh.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013612.1 | NP_001013630.1 | 6 - 7 | 905 | 100 | |
| GenBank mRNA | BC075574.1 | - | 6 - 7 | 905 | 100 | |
| CR855655.2 | - | 6 - 7 | 906 | 100 | ||
More Information
Set Membership: |
|

ugdh.2
Genomic Map