Assay Details
| Gene Symbol: aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015726.1 | NP_001015726.1 | 456 | 57 | ||
| GenBank mRNA | BC089658.1 | - | 456 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aqp7| Gene Symbol: aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015726.1 | NP_001015726.1 | 456 | 57 | ||
| GenBank mRNA | BC089658.1 | - | 456 | 57 | ||
Set Membership: |
|