Assay Details
| Gene Symbol: ttc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015772.1 | NP_001015772.1 | 2 - 3 | 330 | 90 | |
| GenBank mRNA | BC089712.1 | - | 2 - 3 | 330 | 90 | |
More Information
Set Membership: |
|

ttc36
Genomic Map