Assay Details
| Gene Symbol: rasl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015774.1 | NP_001015774.1 | 1 - 2 | 266 | 91 | |
| GenBank mRNA | BC089714.1 | - | 1 - 2 | 266 | 91 | |
More Information
Set Membership: |
|

rasl11b
Genomic Map