Assay Details
| Gene Symbol: cnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015796.1 | NP_001015796.1 | 4 - 5 | 426 | 58 | |
| XM_012953157.2 | XP_012808611.1 | 4 - 5 | 409 | 58 | ||
| GenBank mRNA | BC089746.1 | - | 4 - 5 | 426 | 58 | |
More Information
Set Membership: |
|

cnn2
Genomic Map