Assay Details
| Gene Symbol: gpaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015809.1 | NP_001015809.1 | 12 - 13 | 1652 | 53 | |
| GenBank mRNA | BC090090.1 | - | 12 - 13 | 1652 | 53 | |
More Information
Set Membership: |
|

gpaa1
Genomic Map