Assay Details
| Gene Symbol: pnliprp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015812.2 | NP_001015812.1 | 4 - 5 | 462 | 56 | |
| XM_012954710.2 | XP_012810164.1 | 5 - 6 | 610 | 56 | ||
| XM_012954711.2 | XP_012810165.1 | 5 - 6 | 634 | 56 | ||
| GenBank mRNA | BC090093.1 | - | 4 - 5 | 465 | 56 | |
More Information
Set Membership: |
|

pnliprp2
Genomic Map