Assay Details
| Gene Symbol: ror2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015834.1 | NP_001015834.1 | 3 - 4 | 867 | 68 | |
| XM_018092983.1 | XP_017948472.1 | 3 - 4 | 513 | 68 | ||
| GenBank mRNA | BC090120.1 | - | 3 - 4 | 867 | 68 | |
More Information
Set Membership: |
|

ror2
Genomic Map