Assay Details
| Gene Symbol: bysl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015838.1 | NP_001015838.1 | 3 - 4 | 576 | 68 | |
| GenBank mRNA | BC090127.1 | - | 3 - 4 | 576 | 68 | |
More Information
Set Membership: |
|

bysl
Genomic Map