Assay Details
| Gene Symbol: tuba3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015841.1 | NP_001015841.1 | 2 - 3 | 253 | 74 | |
| GenBank mRNA | BC090130.1 | - | 2 - 3 | 253 | 74 | |
| CT030272.1 | - | 1 - 2 | 232 | 74 | ||
More Information
Set Membership: |
|

tuba3e
Genomic Map