Assay Details
| Gene Symbol: fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015844.1 | NP_001015844.1 | 1 - 2 | 263 | 64 | |
| GenBank mRNA | BC090133.1 | - | 1 - 2 | 263 | 64 | |
More Information
Set Membership: |
|

fn3k
Genomic Map