Assay Details
| Gene Symbol: mnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015849.1 | NP_001015849.1 | 3 - 4 | 200 | 71 | |
| XM_012961773.1 | XP_012817227.1 | 3 - 4 | 384 | 71 | ||
| GenBank mRNA | BC090139.1 | - | 3 - 4 | 200 | 71 | |
More Information
Set Membership: |
|

mnd1
Genomic Map