Assay Details
| Gene Symbol: dusp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015856.1 | NP_001015856.1 | 1 - 2 | 377 | 54 | |
| GenBank mRNA | BC090366.1 | - | 1 - 2 | 377 | 54 | |
More Information
Set Membership: |
|

dusp5
Genomic Map