Assay Details
| Gene Symbol: cmtm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015861.1 | NP_001015861.1 | 3 - 4 | 459 | 94 | |
| GenBank mRNA | BC090372.1 | - | 3 - 4 | 459 | 94 | |
More Information
Set Membership: |
|

cmtm7
Genomic Map