Assay Details
| Gene Symbol: slc50a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015865.1 | NP_001015865.1 | 4 - 5 | 705 | 69 | |
| GenBank mRNA | BC090379.1 | - | 4 - 5 | 705 | 69 | |
More Information
Set Membership: |
|

slc50a1
Genomic Map