Assay Details
| Gene Symbol: eif4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015909.1 | NP_001015909.1 | 3 - 4 | 223 | 106 | |
| GenBank mRNA | CR926428.1 | - | 3 - 4 | 223 | 106 | |
More Information
Set Membership: |
|

eif4e
Genomic Map