Assay Details
| Gene Symbol: slc18b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015938.1 | NP_001015938.1 | 6 - 7 | 638 | 74 | |
| XM_012962951.2 | XP_012818405.1 | 7 - 8 | 801 | 74 | ||
| GenBank mRNA | BC171089.1 | - | 6 - 7 | 630 | 74 | |
| BC171091.1 | - | 6 - 7 | 630 | 74 | ||
| CR926359.1 | - | 6 - 7 | 638 | 74 | ||
More Information
Set Membership: |
|

slc18b1
Genomic Map