Assay Details
| Gene Symbol: agpat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015964.2 | NP_001015964.1 | 5 - 6 | 604 | 64 | |
| GenBank mRNA | BC157183.1 | - | 5 - 6 | 612 | 64 | |
| CR848431.2 | - | 5 - 6 | 604 | 64 | ||
More Information
Set Membership: |
|

agpat5
Genomic Map