Assay Details
| Gene Symbol: asmtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016011.2 | NP_001016011.1 | 7 - 8 | 555 | 93 | |
| GenBank mRNA | BC121542.1 | - | 6 - 7 | 523 | 93 | |
| CR848221.2 | - | 7 - 8 | 555 | 93 | ||
More Information
Set Membership: |
|

asmtl
Genomic Map