Assay Details
| Gene Symbol: ormdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016075.2 | NP_001016075.1 | 3 - 4 | 352 | 96 | |
| GenBank mRNA | BC170729.1 | - | 3 - 4 | 349 | 96 | |
| BC170832.1 | - | 3 - 4 | 349 | 96 | ||
| CR762230.2 | - | 3 - 4 | 352 | 96 | ||
More Information
Set Membership: |
|

ormdl2
Genomic Map