Assay Details
| Gene Symbol: h2afv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016178.2 | NP_001016178.1 | 4 - 5 | 539 | 77 | |
| GenBank mRNA | BC091605.1 | - | 4 - 5 | 539 | 77 | |
| CR761860.2 | - | 4 - 5 | 382 | 77 | ||
More Information
Set Membership: |
|

h2afv
Genomic Map