Assay Details
| Gene Symbol: syngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016195.2 | NP_001016195.1 | 1 - 2 | 390 | 107 | |
| XM_012961070.2 | XP_012816524.1 | 1 - 2 | 478 | 107 | ||
| GenBank mRNA | BC153693.1 | - | 1 - 2 | 424 | 107 | |
| CR926351.2 | - | 1 - 2 | 390 | 107 | ||
| Gene Symbol: tab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC075573.1 | - | 1 - 2 | 382 | 107 | |
More Information
Set Membership: |
|

syngr1
Genomic Map