Assay Details
| Gene Symbol: camk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016269.2 | NP_001016269.1 | 3 - 4 | 376 | 72 | |
| XM_012961079.2 | XP_012816533.1 | 5 - 6 | 556 | 72 | ||
| XM_012961080.1 | XP_012816534.1 | 5 - 6 | 436 | 72 | ||
| XM_012961081.2 | XP_012816535.1 | 4 - 5 | 484 | 72 | ||
| GenBank mRNA | BC090591.1 | - | 3 - 4 | 376 | 72 | |
| CR761672.1 | - | 5 - 6 | 521 | 72 | ||
More Information
Set Membership: |
|

camk1
Genomic Map