Assay Details
| Gene Symbol: aatf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016308.2 | NP_001016308.1 | 10 - 11 | 1458 | 68 | |
| XM_012956944.2 | XP_012812398.1 | 10 - 11 | 1489 | 68 | ||
| XM_012956945.2 | XP_012812399.1 | 10 - 11 | 1489 | 68 | ||
| XM_012956946.2 | XP_012812400.1 | 10 - 11 | 1487 | 68 | ||
| XM_018091245.1 | XP_017946734.1 | 10 - 11 | 1488 | 68 | ||
| GenBank mRNA | BC166292.1 | - | 10 - 11 | 1421 | 68 | |
| BC170972.1 | - | 10 - 11 | 1451 | 68 | ||
| BC170974.1 | - | 10 - 11 | 1451 | 68 | ||
| CR761495.2 | - | 10 - 11 | 1458 | 68 | ||
More Information
Set Membership: |
|

aatf
Genomic Map