Assay Details
| Gene Symbol: gfod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016311.2 | NP_001016311.1 | 1 - 2 | 615 | 80 | |
| GenBank mRNA | CR761488.2 | - | 1 - 2 | 615 | 80 | |
More Information
Set Membership: |
|

gfod1
Genomic Map