Assay Details
| Gene Symbol: txnl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016338.2 | NP_001016338.1 | 2 - 3 | 300 | 118 | |
| XM_012956958.2 | XP_012812412.1 | 3 - 4 | 403 | 118 | ||
| XM_012956959.1 | XP_012812413.1 | 3 - 4 | 500 | 118 | ||
| XM_018091248.1 | XP_017946737.1 | 3 - 4 | 383 | 118 | ||
| GenBank mRNA | BC167881.1 | - | 2 - 3 | 454 | 118 | |
| CR761402.2 | - | 2 - 3 | 300 | 118 | ||
More Information
Set Membership: |
|

txnl4b
Genomic Map