Assay Details
| Gene Symbol: tbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016339.3 | NP_001016339.1 | 2 - 3 | 460 | 67 | |
| XM_012962993.2 | XP_012818447.1 | 3 - 4 | 588 | 67 | ||
| GenBank mRNA | BC160510.1 | - | 2 - 3 | 369 | 67 | |
| BC171174.1 | - | 2 - 3 | 456 | 67 | ||
| BC171176.1 | - | 2 - 3 | 456 | 67 | ||
| CR761393.2 | - | 2 - 3 | 462 | 67 | ||
More Information
Set Membership: |
|

tbp
Genomic Map