Assay Details
| Gene Symbol: mrps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016340.2 | NP_001016340.1 | 2 - 3 | 118 | 63 | |
| XM_012969833.1 | XP_012825287.1 | 2 - 3 | 173 | 63 | ||
| GenBank mRNA | BC157196.1 | - | 1 - 2 | 103 | 63 | |
| CR761392.2 | - | 2 - 3 | 118 | 63 | ||
More Information
Set Membership: |
|

mrps21
Genomic Map