Assay Details
Gene Symbol: celf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016491.2 | NP_001016491.1 | 484 | 105 | ||
GenBank mRNA | BC154063.1 | - | 507 | 105 | ||
BC170955.1 | - | 185 | 105 | |||
CR760829.2 | - | 484 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
