Assay Details
| Gene Symbol: psmb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016503.2 | NP_001016503.1 | 3 - 4 | 316 | 71 | |
| XM_012971771.2 | XP_012827225.1 | 3 - 4 | 473 | 71 | ||
| GenBank mRNA | BC157194.1 | - | 3 - 4 | 324 | 71 | |
| CR760794.2 | - | 3 - 4 | 316 | 71 | ||
More Information
Set Membership: |
|

psmb3
Genomic Map