Assay Details
| Gene Symbol: atp5f1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016516.2 | NP_001016516.1 | 2 - 3 | 300 | 81 | |
| GenBank mRNA | BC135474.1 | - | 2 - 3 | 293 | 81 | |
| CR926220.2 | - | 2 - 3 | 300 | 81 | ||
More Information
Set Membership: |
|

atp5f1d
Genomic Map