Assay Details
| Gene Symbol: kctd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016558.1 | NP_001016558.1 | 3 - 4 | 484 | 104 | |
| XM_012962508.2 | XP_012817962.1 | 4 - 5 | 640 | 104 | ||
| GenBank mRNA | CR760702.1 | - | 3 - 4 | 484 | 104 | |
More Information
Set Membership: |
|

kctd10
Genomic Map