Assay Details
| Gene Symbol: ccdc127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016568.2 | NP_001016568.1 | 1 - 2 | 129 | 92 | |
| GenBank mRNA | BC135132.1 | - | 2 - 3 | 140 | 92 | |
| CR760672.2 | - | 1 - 2 | 129 | 92 | ||
More Information
Set Membership: |
|

ccdc127
Genomic Map