Assay Details
| Gene Symbol: ppp1r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016643.2 | NP_001016643.1 | 5 - 6 | 665 | 68 | |
| XM_012964715.2 | XP_012820169.1 | 5 - 6 | 714 | 68 | ||
| GenBank mRNA | BC121871.1 | - | 4 - 5 | 555 | 68 | |
| CR760399.2 | - | 5 - 6 | 665 | 68 | ||
More Information
Set Membership: |
|

ppp1r42
Genomic Map